Learn More
9-Nitroanthracene matrix substance for MALDI-MS, ≥98.5% (HPLC), MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 56229100MG
 View more versions of this product
Catalog No. 11-101-4848
Add to cart
Description
Description
Specifications
Specifications
602-60-8
MFCD00001248
≥98.5% (HPLC)
141°C to 144°C; 144°C to 150°C
Organic Standard
C14H9 NO2
223.23
Matrix Substance for MALDI-MS
100 mg