Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Nitroanthracene matrix substance for MALDI-MS, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

$148.10 - $1100.00

Specifications

CAS 602-60-8
Percent Purity ≥98.5% (HPLC)
Grade Matrix Substance for MALDI-MS
Linear Formula C14H9 NO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4848
View Documents
MilliporeSigma Supelco
56229100MG
100 mg
Each for $148.10
Add to cart
 
11-101-4847
View Documents
MilliporeSigma Supelco
562291G
1 g
Each for $1,100.00
Add to cart
 
Description

Description

Specifications

Specifications

602-60-8
≥98.5% (HPLC)
Matrix Substance for MALDI-MS
C14H9 NO2
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit