Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Phenanthrol, 60%, Tech., ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
484-17-3
C14H10O
194.233
MFCD00082298
DZKIUEHLEXLYKM-UHFFFAOYSA-N
9-phenanthrol,9-phenanthrenol,9-hydroxyphenanthrene,unii-9fyu45ov9h,ccris 1840,9fyu45ov9h,9-hydroxy-phenanthrene,d0g2mu,acmc-1ad67,dsstox_cid_27592
10229
CHEBI:28820
phenanthren-9-ol
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)O
484-17-3
C14H10O
194.233
MFCD00082298
DZKIUEHLEXLYKM-UHFFFAOYSA-N
9-phenanthrol,9-phenanthrenol,9-hydroxyphenanthrene,unii-9fyu45ov9h,ccris 1840,9fyu45ov9h,9-hydroxy-phenanthrene,d0g2mu,acmc-1ad67,dsstox_cid_27592
10229
CHEBI:28820
phenanthren-9-ol
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Brown
CAS Min % 55.0
CAS Max % 100.0
Assay Percent Range 60%
Beilstein 06, 706
Formula Weight 194.23
Physical Form Powder
Percent Purity ≥55%
Grade Technical
Chemical Name or Material 9-Phenanthrol
Show More Show Less