Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Phenylanthracene, 98%

Click to view available options
Quantity:
1g
5g
602-55-1
C20H14
254.332
MFCD00001252
LUBXLGUQZVKOFP-UHFFFAOYSA-N
anthracene, 9-phenyl,9-phenyl-anthracene,10-phenylanthracene,9-phenyl-anthracen,9-phenylanthracene,acmc-1b14f,anthracene, 9-phenyl-8ci 9ci
11766
9-phenylanthracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=CC4=CC=CC=C42
602-55-1
C20H14
254.332
MFCD00001252
LUBXLGUQZVKOFP-UHFFFAOYSA-N
anthracene, 9-phenyl,9-phenyl-anthracene,10-phenylanthracene,9-phenyl-anthracen,9-phenylanthracene,acmc-1b14f,anthracene, 9-phenyl-8ci 9ci
11766
9-phenylanthracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=CC4=CC=CC=C42
Sensitivity Light Sensitive
Quantity 5g
Color White to Brown
Melting Point 153°C to 155°C (literature)
Boiling Point 417°C (literature)
UN Number UN3077
Beilstein 1910570
Formula Weight 254.33
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 9-Phenylanthracene
Show More Show Less

DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.