Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 9-(Trimethylsilyl)fluorene, 99%
Click to view available options
Quantity:
1g
5g
Specifications
Formula Weight | 238.4 |
Percent Purity | 99% |
Melting Point | 95°C to 98°C |
Quantity | 1g |
Chemical Name or Material | 9-(Trimethylsilyl)fluorene |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures