Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 9-Vinylanthracene, 97%

Click to view available options
Quantity:
1g
5g
2444-68-0
C16H12
204.272
MFCD00003576
OGOYZCQQQFAGRI-UHFFFAOYSA-N
9-vinylanthracene,anthracene, 9-ethenyl,unii-a3692nge8j,9-vinyl-anthracene,9-ethenyl anthracene,9-ethenylanthracen,10-vinylanthracene
17125
9-ethenylanthracene
C=CC1=C2C=CC=CC2=CC3=CC=CC=C31
2444-68-0
C16H12
204.272
MFCD00003576
OGOYZCQQQFAGRI-UHFFFAOYSA-N
9-vinylanthracene,anthracene, 9-ethenyl,unii-a3692nge8j,9-vinyl-anthracene,9-ethenyl anthracene,9-ethenylanthracen,10-vinylanthracene
17125
9-ethenylanthracene
C=CC1=C2C=CC=CC2=CC3=CC=CC=C31
Melting Point 62°C to 65°C
Quantity 5g
Boiling Point 61°C to 66°C (10mmHg)
Beilstein 2041641
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 9-Vinylanthracene

Recommended Storage : 2° to 8°C