Learn More
9-Vinylcarbazole, 98+%, Thermo Scientific Chemicals
Description
9-Vinylcarbazole is used as a monomer in the production of poly(vinylcarbazole), a conductive polymer, in which conductivity is photon-dependent. It is also used in the photoreceptors of photocopiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
9-Vinylcarbazole is used as a monomer in the production of poly(vinylcarbazole), a conductive polymer, in which conductivity is photon-dependent. It is also used in the photoreceptors of photocopiers.
Solubility
Soluble in acetonitrile.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
Melting Point | 60°C to 65°C |
Boiling Point | 154°C to 155°C |
Quantity | 100 g |
UN Number | UN2811 |
Beilstein | 132988 |
Solubility Information | Soluble in acetonitrile. |
Formula Weight | 193.24 |
Percent Purity | ≥98% |
Chemical Name or Material | 9-Vinylcarbazole |
Safety and Handling
Hazard Category | H300-H312-H315-H317-H319-H341 |
Hazard Statement | GHS H Statement H341-H302-H312-H315-H317 Suspected of causing genetic defects. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P261-P264b-P270-P272-P280-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 216-055-0 |
RTECSNumber | FE6350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.