Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 96-Well Advanced TC™ Treated µClear™ F-Bottom/Chimney Well Microplates

For cultivation of fastidious and sensitive cells. Greiner Bio-One 96-Well Advanced TC™ Treated µClear™ F-Bottom/Chimney Well Microplates enhances cell attachment.

$450.50

Specifications

Autoclavable Not Autoclavable
Lid With Lid
Bottom Flat
Culture Area 34 mm2
Dimensions (L x W x H) 128 x 85 x 14 mm
View More Specs

Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
07-000-172
View Documents
Greiner Bio-One
655983
White
Case of 32 for $450.50
Add to cart
 
Description

Description

Advanced TC polymer modification changes plastic surface in a way that increases primary and long-term cell adhesion and positively influences cell-dependant features and function enhanced cell attachment and higher proliferation rates facilitate rapid cell expansion even under restricted growth conditions.

  • Provide conditions for improved cell adherence, accelerated cell expansion and increase overall cell yield
  • For use with serum reduced and serum-free media
  • Allows differentiation of semi-adherent cells
  • Non-pyrogenic and non-cytotoxic
  • Homogenous cell growth
  • Higher transgene activity
  • µClear coated well bottom
Specifications

Specifications

Not Autoclavable
Flat
128 x 85 x 14 mm
Sterile
With Lid
34 mm2
Polystyrene
Advanced TC
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit