Learn More
Greiner Bio-One 96-Well Chimney Designed No-Bottom Microplates
Ideal for work with nonpolar proteins and peptides. Greiner Bio-One Polystyrene 96 Well No-Bottom Microplate features chimney-well design to help prevent cross-contamination.
$220.00 - $234.00
Specifications
Bottom No Bottom
Well Design Chimney
Material Polystyrene
Format Standard
View More Specs
Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
07-000-998
View Documents
Greiner Bio-One
65500005
White
Case of 40 for $220.00
Add to cart
07-000-999
View Documents
Greiner Bio-One
65500006
Black
Case of 40 for $234.00
Add to cart
Description
Description
  • Virgin polystyrene construction
  • Medium binding surface
  • Designed without bottom
  • Features well diameter design that offers one-half the well surface area of standard plates
  • Allows 50% sample volume reduction without m
Specifications
Specifications
No Bottom
Chimney
Polystyrene
Standard