Learn More
Greiner Bio-One 96-Well Chimney Designed No-Bottom Microplates
Ideal for work with nonpolar proteins and peptides. Greiner Bio-One Polystyrene 96 Well No-Bottom Microplate features chimney-well design to help prevent cross-contamination.
Manufacturer:  Greiner Bio-One 65500005
 View more versions of this product
Catalog No. 07-000-998
Add to cart
Description
Description
  • Virgin polystyrene construction
  • Medium binding surface
  • Designed without bottom
  • Features well diameter design that offers one-half the well surface area of standard plates
  • Allows 50% sample volume reduction without m
Specifications
Specifications
No bottom
96 well
3000 pieces/case
Microplate
White
Polystyrene
Chimney