Learn More
Thermo Scientific™ 96-Well Flipper™ Microtube Racks
Protect samples and allow racks to be stacked using this 96-Well Flipper Microtube Rack.
Manufacturer:  Thermo Scientific™ 8780
 View more versions of this product
Catalog No. 21-402-20
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Protect samples and allow racks to be stacked using the Thermo Scientific? 96-Well Flipper? Microtube Rack.
  • Holds 0.5mL tubes on one side, 1.5mL tubes on the other
  • Molded with an alphanumeric grid for easy tube identification
  • Several colors available
Description & Specifications
Specifications
Fluorescent Green
0.5mL Tubes,1.5mL Tubes,2.0 mL Tubes
10 racks
Flipper Rack
Polypropylene
96-Well Flipper Rack