Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One 96-well Non-treated Polypropylene Microplates
Features high chemical resistance. Greiner Bio-One 96-Well Non-treated Polypropylene Microplates is free of detectable DNase, Rnase and human DNA.
Manufacturer: Greiner Bio-One 655205
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Polypropylene
- High chemical resistance
- Sealable with adhesive films and heat sealer
- Can be traced back to production through defined LOT nu
Certifications
DIN ISO 9001
Specifications
Flat | |
Without Lid | |
Inner bags of 10 | |
Polypropylene | |
Non-sterile | |
100/Cs. |
Standard | |
8 x 12 | |
Green | |
96 | |
Non-treated | |
Chimney |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title