Learn More
Greiner Bio-One 96-well Non-treated Polypropylene Microplates
Features high chemical resistance. Greiner Bio-One 96-Well Non-treated Polypropylene Microplates is free of detectable DNase, Rnase and human DNA.
Manufacturer: Greiner Bio-One 655206
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Polypropylene
- High chemical resistance
- Sealable with adhesive films and heat sealer
- Can be traced back to production through defined LOT nu
Certifications
DIN ISO 9001
Specifications
Flat | |
Without Lid | |
Yellow | |
96 | |
Non-treated | |
Chimney |
Standard | |
8 x 12 | |
Polypropylene | |
Non-sterile | |
100/Cs. |