Learn More
Greiner Bio-One 96-Well Non-treated Polystyrene Microplates
For agglutination tests. Greiner Bio-One 96-Well Non-treated Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
$280.00 - $1522.00
Specifications
Material Polystyrene
Lid Without Lid
Format Standard
Color Clear
View More Specs
Products 5
Catalog Number Mfr. No. Bottom Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Bottom Sterility Price Quantity & Availability  
07-000-124
View Documents
Greiner Bio-One
655101
Flat Non-sterile
Case of 100 for $289.50
Add to cart
07-000-140
View Documents
Greiner Bio-One
651101
V Non-sterile
Case of 100 for $280.00
Add to cart
07-000-609
View Documents
Greiner Bio-One
650101
U Non-sterile
Case of 100 for $282.50
Add to cart
07-000-610
View Documents
Greiner Bio-One
650162
U Sterile
Case of 375 for $1,522.00
Add to cart
07-000-611
View Documents
Greiner Bio-One
650172
U Non-sterile This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Non-Pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Material: Polystyrene
  • Can be traced all the way back to production through a defined LOT number system
  • No sharp corners, can be pipetted easily and cleanlyCertifications
    DIN ISO 9001
Specifications
Specifications
Polystyrene
Without Lid
Standard
Clear