Learn More
Eppendorf™ 96-Well Protein LoBind Deep Well Plates
Use these plates in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results. Eppendorf™ Deepwell Protein LoBind Plates recover significantly more protein for downstream analyses and enzymes remain active.
Specifications
Format Standard
Array 8 x 12
No. of Wells 96
Dnase Free Yes
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Well Color Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Well Color Packaging Price Quantity & Availability  
E0030504305
View Documents
Eppendorf™
0030504305
2000 μL White 20 Plates (5 Bags x 4 Plates)
Pack of 1 for $510.00
N/A
Edge
Description
Description
  • LoBind material guarantees maximum sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: of human DNA, DNAase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Unique OptiTrack™ matrix: 30% faster sample identification and fewer pipetting errors
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface guarantee reliable sealing in plates
Specifications
Specifications
Standard
8 x 12
96
Yes