Learn More
Thermo Scientific™ 96-Well Semi-Skirted PCR Plates
Break plates apart into 24- or 48-well formats with the special design of these 96-well semi-skirted PCR plates, made from 100% pure polypropylene.
Manufacturer:  Thermo Scientific™ 3484L
 View more versions of this product
Catalog No. 21-402-451
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Break plates apart into 24- or 48-well formats with the special design of these Thermo Scientific? 96-well semi-skirted PCR plates, made from 100% pure polypropylene.
  • Thin-wall design for optimal heat transfer
  • Elevated rims for secure sealing
Description & Specifications
Specifications
Black
Polypropylene
Semi-skirted
300µL