Thermo Scientific™ 96-Well Skirted PCR Plates

Ensure uniform sealing with these PCR plates, molded from 100% virgin polypropylene and featuring elevated rims.

Manufacturer: Thermo Scientific™ 3482L

 View more versions of this product

Catalog No. 21-402-443

  Check Availability
Add to Cart

    Specifications

    Barcode No Barcode
    For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
    Format 96-well plate|Skirted
    Skirt Style Skirted
    Material Polypropylene,Polypropylene
    Color Black
    Description Skirted

    Ensure uniform sealing with Thermo Scientific™ PCR plates, molded from 100% virgin polypropylene and featuring elevated rims.

    • Thermo Scientific™ PurePak™ packaging reduces contamination from reaching into a bulk bag of tubes by dividing tubes into ten separate PurePaks
    • Certified Rnase-, DNase- and pyrogen-free

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit