Learn More
Axygen™ 96-well tips, Clear
Axygen™ Maxymum Recovery™ tips have been developed to work with the Agilent/Velocity11 and MGI Tech automation systems, which can be enabled for the high throughput of magnetic bead based RNA extraction of biological samples or pre-configured and qualified to automate high throughput pre-PCR samples.
$70.83 - $929.00
Specifications
Color | Clear |
---|---|
Format | Rack, 96-well |
Quantity | 96 Tips/Rk, 10 Racks/Pk., 5 Pk./Cs |
For Use With (Equipment) | Agilent Bravo and MGI Tech MGISP Automation Systems |
Material | Polypropylene |
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Filtered | Tip Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Filtered | Tip Style | Price | Quantity & Availability | ||
14-222-377
|
Axygen™
VTF205RS |
200 μL | Sterile | Filtered | Standard |
Case
for $70.83
Save 90% Reg : |
|
14-222-378
|
Axygen™
VTF205LRS |
200 μL | Sterile | Filtered | Low Retention, Maxymum Recovery |
Case for $929.00
|
|
14-222-376
|
Axygen™
VT255R |
250 μL | Sterile | Non-Filtered | Standard |
Case for $365.00
|
|
14-222-380
|
Axygen™
VT255LR |
250 μL | Non-sterile | Non-Filtered | Low Retention, Maxymum Recovery |
Case for $416.00
|
|
14-222-384
|
Axygen™
VT255RS |
250 μL | Sterile | Non-Filtered | Standard |
Case of 4800 for $400.50
|
|
14-222-379
|
Axygen™
VT255LRS |
250 μL | Sterile | Non-Filtered | Low Retention, Maxymum Recovery |
Case for $592.00
|
|
Description
- Tip format: 96 tips
- Tip volume range: Up to 250 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- non-sterile or sterile
- Axygen Maxymum Recovery surface
- Compatibility:
- Bravo automated liquid handling platform
- VPrep™ vertical liquid handling pipetting station
- MGISP-960
- MGISP-100
Specifications
Clear | |
96 Tips/Rk, 10 Racks/Pk., 5 Pk./Cs | |
Polypropylene |
Rack, 96-well | |
Agilent Bravo and MGI Tech MGISP Automation Systems |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.