Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ 96-Well twin.tec™ PCR Plates, Forensic DNA grade Available on GSA/VA Contract for Federal Government customers only.

Reduces the risk of cross contamination with raised rims for effective sealing. Eppendorf™ 96-Well twin.tec™ PCR Plates benefits in automation and high throughput processes.

$64.30

Specifications

Type Forensic DNA Grade
Rimmed Yes
For Use With (Application) PCR with medium to high sample throughput
Color Clear
Format Standard
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) Well Skirt Style Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Well Skirt Style Price Quantity & Availability  
E0030129610 Available on GSA/VA Contract for Federal Government customers only. View Documents Eppendorf™
0030129610
250 μL Semi Each for $64.30
Add to cart
 
Description

Description

  • Combines polycarbonate frame and polypropylene wells for optimum performance
  • Thin-walled polypropylene wells offer optimum heat transfer to sample
  • Solid and torque-resistant polycarbonate frame
  • Improved well-to-well tolerance
  • Cut-away corner and alphanumeric grid-referencing
  • Raised well rims for effective sealing
  • Free of any detectable human DNA, DNase, RNase and PCR inhibitors
  • Ideal for quantitative real-time PCR

Applications

  • Standard PCR
  • Real-time PCR
  • High Throughput PCR
  • Small Volume Sample Handling

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Forensic DNA Grade
PCR with medium to high sample throughput
Standard
Polycarbonate Frame, Polypropylene Wells
Yes
Clear
10
Not Autoclavable
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit