Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ 96-Well Twin.tec™ PCR Plates, Skirted
GSA_VA

Skirted 96-Well Twin.tec™ PCR Plates are ideal for extreme applications such as multiple heat sealing.

$195.56

Specifications

Certifications/Compliance RNase, DNase free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Product Type PCR Plate 96
View More Specs

Products 5
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
E951020401
GSA_VA
View Documents
Eppendorf™
951020401
Clear
Case of 25 for $195.56
Add to cart
 
E951020427
View Documents
Eppendorf™
951020427
Yellow
Case of 25 for $195.56
Add to cart
 
E951020443
GSA_VA
View Documents
Eppendorf™
951020443
Green
Case of 25 for $195.56
Add to cart
 
E951020460
GSA_VA
View Documents
Eppendorf™
951020460
Blue
Case of 25 for $195.56
Add to cart
 
E951020486
View Documents
Eppendorf™
951020486
Red
Case of 25 for $195.56
Add to cart
 
Description

Description

  • Optimized rim for barcodes and automated work processes
  • Within SBS footprint recommendations (127.76 × 85.47 mm ±0.25 mm)
  • Stackable
  • Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
  • Low profile design enhances efficiency of PCR and enables the highest efficiency for small sample volumes
  • 150 µL max. well volume (when used with cap strips)
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well – OptiTrack™ matrix for faster sample identification and fewer pipetting errors

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Certifications

Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.

Specifications

Specifications

RNase, DNase free
8 x 12
PCR Plate 96
Polycarbonate
96
25
25/Pk.
Standard
Autoclavable
Skirted
150 μL
Ideal for extreme applications such as multiple heat sealing.
PCR Clean
Colorless
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit