Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ 96-Well twin.tec™ real-time PCR Plates Available on GSA/VA Contract for Federal Government customers only.

Increases the reflection of fluorescence in real-time PCR and other fluorescence assays due to white wells.

$90.00

Specifications

Type Forensic DNA Grade
Rimmed Yes
No. of Wells 96
For Use With (Application) Real-time PCR
Color White
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Well Skirt Style Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Well Skirt Style Price Quantity & Availability  
E0030129636 Available on GSA/VA Contract for Federal Government customers only. View Documents Eppendorf™
0030129636
150 μL Skirted Each for $90.00
Add to cart
 
E0030129644 Available on GSA/VA Contract for Federal Government customers only. View Documents Eppendorf™
0030129644
250 μL Semi Each for $90.00
Add to cart
 
Description

Description

  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121°C, 20min)

Applications

  • Real-time PCR
  • Real-time PCR
  • Melting Curve Analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Forensic DNA Grade
96
White
10
Autoclavable
Yes
Real-time PCR
Standard
Polypropylene
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit