Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One 96-Well Conical Bottom (V) Polypropylene Microplates
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Conical Bottom (V) Polypropylene Microplates is free of detectable DNase, RNase, human DNA.
Manufacturer: Greiner Bio-One 651206
Description
- Uniform external dimensions
- Well to well spacing of 9mm
- Features alphanumeric coding
- Sealable with adhesive films and heat seal
Specifications
Not Autoclavable | |
Without Lid | |
Free of detectable DNase/RNase, human DNA and pyrogens. | |
128 x 85 x 15 mm | |
100/Cs. | |
96 |
Standard | |
V | |
Yellow | |
Polypropylene | |
Non-sterile | |
8 x 12 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title