Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9h-fluorene-1-carboxylic Acid, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
6276-04-6
C6H3IN2O4
294.00
MFCD00001130
AISNAASNOWRWIR-UHFFFAOYSA-N
3,5-dinitroiodobenzene,benzene, 1-iodo-3,5-dinitro,acmc-20a9ow,1,3-dinitro-5-iodobenzene,#,benzene,1-iodo-3,5-dinitro,1-iodo-3,5-dinitrobenzene
235269
1-iodo-3,5-dinitrobenzene
C1=C(C=C(C=C1[N+](=O)[O-])I)[N+](=O)[O-]
6276-04-6
C6H3IN2O4
294.00
MFCD00001130
AISNAASNOWRWIR-UHFFFAOYSA-N
3,5-dinitroiodobenzene,benzene, 1-iodo-3,5-dinitro,acmc-20a9ow,1,3-dinitro-5-iodobenzene,#,benzene,1-iodo-3,5-dinitro,1-iodo-3,5-dinitrobenzene
235269
1-iodo-3,5-dinitrobenzene
C1=C(C=C(C=C1[N+](=O)[O-])I)[N+](=O)[O-]
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 10g
Color Brown
Melting Point 246°C to 250°C
Beilstein 09, 690
Formula Weight 210.23
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 9H-Fluorene-1-carboxylic acid

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.