Fisherbrand™ Vials and Closures Available on GSA/VA Contract for Federal Government customers only.

Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear or amber glass vials and 100 screw thread caps with pre-assembled septa.

 View more versions of this product

Catalog No. 13-622-189

  Check Availability
Add to Cart

    Specifications

    Septa Red PTFE/White Silicone/Red PTFE
    For Use With Chromatography
    Material Glass
    Color Amber
    Patched Yes
    No. per Pack 100
    Includes Vials and Closures
    Type Autosampler Vial
    Vial Closure Size 9mm
    Closure Color Blue
    Closure Type Screw
    Description Amber Glass w/I-D Patch, Red PTFE/White Silicone/Red PTFE septum, Blue DP Cap
    Item Description Assembled Kit

    Fisherbrand™ Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.

    Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa.

     
    Through its distinct features, this convenience kit allows you to:
     
    · Save time during sample preparation
    · Reduce the risk of contamination
    · Unassembled kits
    –– Include 100 vials and 100 caps with pre-assembled septa
    –– Reusable two compartment trays protect vials and closures while keeping matching supplies together
    –– Clear trays make it easy to keep track of available supplies without opening containers
    · Assembled kits
    –– Include 100 vials with pre-attached caps and septa
    –– Packaged in convenient vial trays with clear covers

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit