Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Caps with FEP/Natural Rubber Septa

Available in multiple colors and package sizes.

Manufacturer:  DWK Life Sciences 090020KT

 View more versions of this product

Catalog No. 15-111-024



Description

Description

Specifications

Specifications

Black
No
FEP/Natural Rubber/FEP
Polypropylene
100
9 mm Autosampler Vials - MicroLiter
FEP/Natural Rubber
9 mm ScrewTin Cap
Screw Thread
9 mm
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit