Thermo Scientific 9mm Certified Amber Screw Thread Kits
Use certified clean, low particle, low background vials for you high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
Manufacturer: Thermo Scientific CERT4000-72LV
 View more versions of this product
Catalog No. 03-377-343
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Type Unassembled Vial Kit
    Vial Closure Size 9-425mm
    Closure Material Bonded PTFE/Silicone
    Closure Type Screw Thread
    Septa PTFE/Silicone
    Material Glass
    Color Amber
    Patched Yes
    No. per Pack 100
    Vial Volume (Metric) 350µL
    Item Description Convenience Kit, Certified 9mm Screw Vial, 350µL Fused Insert
    Description Certified Target DP 350µL Amber Glass I-D Fused Insert Vial, 9mm Screw Thread
    Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific™ 9mm Amber Glass Screw MS Certified Vial Convenience Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.
    Unassembled kits
    • 100 vials and 100 caps with pre-assembled septa 
    • Reusable two-compartment trays protect vials and closures while keeping matching supplies together 
    • Clear trays make it easy to keep track of available supplies without opening containers
    Assembled kits
    • 100 vials with pre-attached caps and septa 
    • Packaged in convenient vial trays with clear covers
  • Description & Specifications
Related Products
Product Content Feedback