Learn More
Thermo Scientific 9mm Certified Clear Screw Thread Kits
Use certified clean, low particle, low background vials for you high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
Manufacturer:  Thermo Scientific CERT4000-69LV
 View more versions of this product
Catalog No. 03-377-339
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific? National 9mm Clear Glass Screw Thread Certified Vial Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.
Unassembled kits
  • 100 vials and 100 caps with pre-assembled septa 
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together 
  • Clear trays make it easy to keep track of available supplies without opening containers
Assembled kits
  • 100 vials with pre-attached caps and septa 
  • Packaged in convenient vial trays with clear covers
Description & Specifications
Specifications
Unassembled Vial Kit
350µL
Screw Thread
Certified Target DP 350µL Clear Glass I-D Fused Insert Vial, 9mm Screw Thread
Glass
Yes
9-425mm
Bonded PTFE/Silicone
Convenience Kit, Certified 9mm Screw Vial, 350µL Fused Insert
PTFE/Silicone
Clear
100