Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Thermo Scientific™ 9mm MS Certified Amber Wide Opening Screw Thread Kits
Use certified clean, low particle, low background vials for you high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
Manufacturer: Thermo Scientific™ MSCERT4000-35W
 View more versions of this product
Catalog No. 03-394-205
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Related Products
Product Content Feedback