Learn More
Thermo Scientific™ Convenience Kit, 9mm SureStop Screw Vial, 2mL with AVCS Closure - PROMO
Use certified clean, low particle, low background vials for you high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
Specifications
Material Glass, PTFE, Silicone
Color Clear
Closure Color Blue
No. per Pack 100
View More Specs
Products 2
Catalog Number Mfr. No. Septum Pre-slit Price Quantity & Availability  
Catalog Number Mfr. No. Septum Pre-slit Price Quantity & Availability  
03-377-300PM
View Documents
Thermo Scientific™
PROMO/MSCERT500034W
Blue Silicone/PTFE No This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-377-306PM
View Documents
Thermo Scientific™
PROMO/MSCERT500040W
Blue Silicone/PTFE Yes This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific™ National 9mm Clear Glass Screw Thread Certified Vial Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.
Unassembled kits
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers
Assembled kits
  • 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers
Specifications
Specifications
Glass, PTFE, Silicone
Clear
Blue
100
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.