Thermo Scientific 9mm MS Certified Clear Screw Thread Kits
Use certified clean, low particle, low background vials for you high sensitivity LC/MS and GC/MS applications. They are fully lot-tested for 15 critical parameters.
Manufacturer: Thermo Scientific MSCERT4000-37LVW
 View all options for this product
Catalog No. 03-394-207
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Item Description Convenience Kit, 9mm 350µL Fused Insert Screw Vial
    Material Glass
    Color Clear
    Patched Yes
    No. per Pack 100
    Use certified clean, low particle, low background vials for your LC/MS, GC/MS and chromatography high sensitivity applications. Thermo Scientific™ National 9mm Clear Glass Screw Thread Certified Vial Kits are fully lot-tested by multiple methods for 15 critical parameters. Every pack comes with a Certificate of Conformance. Manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass, vials are available in assembled and unassembled kits. An assortment of wide opening 9mm vials and a selection of closures are available to suit your application.
    Unassembled kits
    • 100 vials and 100 caps with pre-assembled septa 
    • Reusable two-compartment trays protect vials and closures while keeping matching supplies together 
    • Clear trays make it easy to keep track of available supplies without opening containers
    Assembled kits
    • 100 vials with pre-attached caps and septa 
    • Packaged in convenient vial trays with clear covers
  • Description & Specifications
Related Products
Product Content Feedback