Learn More
Thermo Scientific™ 9mm Non-Assembled Clear Screw Thread Wide Opening Autosampler Vial Kit PROMO
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa.
Manufacturer:  Thermo Scientific™ C5000586WPR
 View more versions of this product
Catalog No. 03-376-607PM
  Check Availability
Add to cart
Description
Description
Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 9mm Wide Opening Clear Vial Convenience kits (Unassembled). Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers. Choose from Thermo Scientific™™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Specifications
Specifications
Clear Glass
Red, White
SureStop Convenience Kit
Red PTFE/White Silicone/Red PTFE Septum
Clear
100
2mL
PTFE; Silicone
2mL, Clear Glass w/ID Patch, with Red PTFE/White Silicone/Red PTFE Septum
Glass
Yes
Documents
Promotions
Promotions
  • Thermo Scientific
    Get 25% off Thermo Scientific™ SureStop™ Vial Kits. Consistent seal eliminates user-to-user variation.
    09-30-2018
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.