Thermo Scientific 9mm Non-Assembled Clear Screw Thread Wide Opening Autosampler Vial Kits

Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa.

Manufacturer: Thermo Scientific C400093W

 View more versions of this product

Catalog No. 03-377-293

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    No. per Pack 100
    Type Assembled Vial Kit
    Vial Volume (Metric) 2mL
    Closure Color Gray
    Description Clear Glass I-D; Bonded PTFE/Silicone, Pre-slit septum
    Septa Bonded PTFE/Silicone, Pre-slit
    Material Glass
    Color Clear
    Patched Yes
    Vial Closure Size 9-425mm
    Item Description Convenience Kit

    Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 9mm Wide Opening Clear Vial Convenience kits (Unassembled). Each kit includes 100 clear glass vials and 100 screw thread caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.

    • Convenience Kits with 2mL, 12 x 32mm vials
    • I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
    • Blue caps with your choice of vial and preassembled seals with septa
    • Packaged in two-compartment benchtop storage tray
    • All items remain dust-free
    • Caps with pre-inserted septa are not attached to vials (packed in separate polybag)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit