Learn More
Epredia™ HistoScreen™ Cassettes in Tube Packs
Eliminates the need for sponges or bags, ideal for very small biopsy tissue samples. Cassettes are compatible with Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Supplier: Epredia™ B851001OR
This item is not returnable.
View return policy
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Mesh | |
750/Cs. | |
Yes | |
For Use With PrintMate™ Printers |
Yes | |
Separate Lid, Back Hinge | |
Orange | |
HistoScreen Cassette |