Learn More
Beckman Coulter Suitability Standard Solution Kit
Supplier: Beckman Coulter FG7018402
PAT700 System Suitability Standards Kit, includes: 0.500 mg C/L sucrose, reagent water and USP 1,4-benzoquinone solution in RFID bottles.