Learn More
Dwyer Instruments Inc Flow Meter, 5"""" Scale, 1-10 SCFH Air, SS Valve
SDP
Supplier:  Dwyer Instruments Inc RMB50SSV
Encompass_Preferred
Accurate within ±3% of full scale reading. Scales are brushed aluminum, coated with epoxy and the graduations are on both sides of the indicating tube.
Catalog No. NC1350892