Learn More
Tri Continent Scientific 5 mL PTFE 3cmSyringe, 25/pack
Supplier:  Tri Continent Scientific 10486
Encompass
This 3cm syringe fits TriContinent C-Series Pumps, Tecan's Cavro™ XP3000 and XCalibur. TriContinent Prime Series Syringes can be summarized in two words: quality and customization. For over four decades, we have been producing syringes of the finest quality, specially designed for use with syringe pumps. Each of our Prime Series Syringes is made from premium borosilicate glass, and has precision bore diameters of +/- 0.0002". Our syringe seal materials include high-quality PTFE, UHMWPE, or custom UHMWPE blends.
Catalog No. NC1368559
Due to product restrictions, please to purchase or view availability for this product.