Learn More
ACROBiosystems Recombinant Protein;Human TIM-3 / HAVCR2 (22-200) Protein, Fc Tag;HEK293;100UG;TM3,HAVCR2,HAVcr-2,KIM3,TIM3,TIMD-3,TIMD3,Tim-3,FLJ14428,KIM-3,KIM3
Supplier:  ACROBiosystems TM3H5258100UG
Encompass_Preferred
Recombinant Protein;Human TIM-3 / HAVCR2 (22-200) Protein, Fc Tag;100UG;Human TIM-3, Fc Tag (TM3-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.5 kDa. The protein migrates as 55-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIM-3 (22-200), Fc Tag (Cat. No. TM3-H5258) at 2 μg/mL (100 µl/well),can bind anti-TIM3 mAb with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. NC1424209
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.