Learn More
Dwyer Instruments Inc Mark III Handheld Digital Manometer, 0-40""""
SDP
Supplier:  Dwyer Instruments Inc 4752FM
Encompass_Preferred
Monitoring natural gas pressures on boilers and other combustion equipment. Air velocity monitoring, when used with a Dwyer Pitot tube and AV calculator. Field calibration of other instruments. Monitoring or troubleshooting HVAC systems.
Catalog No. NC1464602