Learn More
ACROBiosystems Recombinant Protein;Human CD58 / LFA-3 Protein, Fc Tag ;HEK293;100UG;LF3,CD58,LFA3,Ag3,LFA-3,Ag-3
Supplier: ACROBiosystems LF3H5256100UG
Recombinant Protein;Human CD58 / LFA-3 Protein, Fc Tag ;100UG;Human CD58, Fc Tag (LF3-H5256) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.1 kDa. As a result of differtent glycosylation, the protein migrates as 60-75 kDa under reducing (R) condition, and 125-150 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human CD2, His Tag (Cat. No. CD2-H5226) at 2μg/mL (100 μL/well) can bind Human CD58, Fc Tag (Cat. No. LF3-H5256) with a linear range of 10-156 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.