Learn More
Columbus Chemical, Inc. Hydrochloric Acid, 7647-01-0, MFCD00011324, 1G 4X3.8L
Supplier: Columbus Chemical, Inc. 25750080405
Empirical Formula (Hill Notation): HCl, Molecular Weight: 36.46. Hydrochloric acid has been used: for the solubilization of formazan crystals, to adjust pH in various buffers, for the preparation of giant reed extract, in cell viability assays, for DNA denaturation, and for pH adjustments in sucrose hydrolysis experiment.