Learn More
101 BIO Pure Plasmid 96 Miniprep Kit (4x96 reactions) DFS Item
Manufacturer:  101 BIO W0506496
This kit is based on a modified alkaline lysis method, with improved reagent formulation and silica membrane, which has high adsorption efficiency of the plasmid DNA. The proteins, genomes, RNA and other impurities can be removed after washing and an elution. 3 ~ 10 μg of plasmid DNA can be purified from one well (1 ~ 1.3 mL) of overnight culture of E. Coli in LB medium in 96-well plate with this kit. The extracted DNA can be directly used for PCR, restriction enzyme digestion, and sequencing. No need to precipitate, concentrate or desalt.
Catalog No. NC1492022