Learn More
FUJIFILM IRVINE SCIENTIFIC INC iCell Astrocytes Kit, 01434, 1 Vial: > 1 x 106 cells/vial
Supplier: FUJIFILM IRVINE SCIENTIFIC INC R1092
CDI’s iCell Astrocytes are human iPS cell-derived astrocytes. They provide a readily accessible, consistent, and biologically relevant source of astrocytes for the study of synaptic transmission and plasticity in normal CNS function and disease progression.