Learn More
Wesco Industrial Products Stainless Steel Pallet Truck
Supplier: Wesco Industrial Products 272152
Stainless Steel Pallet Truck 1 year pump warranty. For use in food industry, laboratories and areas where corrosive solutions are needed. 6 1/4" wide forks. 7" nylon steering wheels. 3" nylon load wheels. Nylon entry rollers. 210˚ steering radius. Comfort-grip coated handle. Capacity 5500 lbs Weight 164 lbs. Lowered Height 3", raised height 7".
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.