Learn More
Thermo Scientific™ School COVID-19 Sample Collection Supplies Products for Science Education
Easy and ready-to-use COVID-19 sample collection supplies for the Thermo Fisher Scientific ReadyCheckGo™ Testing Program.
Manufacturer:  Thermo Scientific™ R12720
Catalog No. R12720
Add to cart
This item is not returnable. View return policy
Description
Description
  • Ready-to-use COVID-19 sample collection supplies
  • Viral transport media
  • 50 kits (300 participants) per Pack; one kit equates to a testing pool of 6 participants
Required Accessories: Test Kit 1 (Cat. No. 16-888-100) or Test Kit 2 (16-888-101).
Specifications
Specifications
Kit
Viral Transport Media Kit
Non-sterile
Nasal
Transportation of Viruses
SDS
Documents
Product Certifications
Accessories