Learn More
Edvotek™ Detecting Risk Factors for Alzheimer's Disease Using Western Blot Kit
Helps students to understand the theory and application of western blotting. Students will perform a western blot to determine simulated clinical trial participants' risk of developing Alzheimer's Disease.
Supplier: Edvotek™ 1115
Description
- Storage: Some Components Require Refrigerator Storage
- For 6 Lab Groups, With 2 Groups Sharing a Gel
Specifications
Kit | |
14 to 20 | |
12% Denaturing Polyacrylamide Gels (3), Vertical Gel Electrophoresis Apparatus, Power Supply, Shaker Platform (optional), Automatic Pipettes with Tips, Microtest Tubes, Beakers, Transfer Pipettes, Graduated Cylinders, Plastic Wrap, Scissors, Trays/Containers, Forceps, Paper Towels, Lab Gloves, Safety Goggles, Methanol, and Distilled Water | |
6 Lab Groups |
9 to Undergraduate | |
Instructions, Pre-stained LyphoProteinTM Gel Marker, Pre-stained LyphoProteinsTM, Immunochemical and Blotting Reagents, 10 x Tris-glycine-SDS Chamber Buffer, 10 x Tris-glycine Powdered Transfer Buffer, Practice Gel Loading Solution, Nitrocellulose Membrane, Filter Paper (Small and Large) | |
Some Components Require Refrigerator Storage |