Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Small Group Learning: Science In The Kitchen
This comprehensive kit incorporates a variety of scientific techniques all themed around common kitchen materials.
Supplier: Aldon Corporation IS9355SGL
Description
Students will:
- Perform chromatography on commercial food colors
- Perform chemical tests to identify an unknown cooking ingredient
- Examine the protein digesting ability of a common meat tenderizing enzyme
- Use titration to quantify vitamin C levels
- Test an unknown juice or soda (not provided)
- Examine some of the differences and similarities in the materials used to clean (soap, hand dishwashing detergent, and machine dishwashing detergent)
- Kit contains enough materials for 5 groups
- Teacher's Manual and Student Study Guide copymasters are included
- Aligned to the Next Generation Science Standards (NGSS)
- Disciplinary Core Ideas: PS1.A; PS1.B
- Performance Expectations: MS-PS1-2; MS-PS1-3; HS-PS1-3
- Cross Cutting Concepts: Patterns; Structure and Function
- Engineering Practices: Analyzing and Interpreting Data; Obtaining, Evaluating and Communicating Information; Planning and Carrying Out Investigations
Specifications
Next Gen Aligned | |
White | |
13 to 20 | |
1 x 0.5 mL Food Color, Red; 1 x 0.5 mL Food Color, Blue; 1 x 0.5 mL Food Color, Green; 1 x 0.5 mL Food Color, Yellow; 1 x 10g Baking Powder; 1 x 10g Baking Soda; 1 x 10g Corn Starch; 1 x 10g Unknown Powder; 1 x 10 mL Acetic Acid, 5% Solution; 1 x 10 mL Iodine Solution; 1 x 5g Papain; 1 x 5g Gelatin; 1 capsule Ascorbic Acid; 1 x 30 mL Iodine | |
1 | |
11.75 x 9 x 4 in. |
Education Kit | |
7 to 14 | |
Consumer Chemistry | |
Cardboard Box Kit | |
2.8 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Next Generation Science Standards is a registered trademark of Achieve. Neither Achieve nor the lead states and partners that developed the Next Generation Science Standards was involved in the production of, and does not endorse, this product.
Spot an opportunity for improvement?
Provide Content Correction