Learn More
Motic Swift Line Student Monocular LED Microscope M3604-4 Products for Science Education
Built-in handle, variable LED illumination and lead-free objectives are just some of the state-of-the-art features that makes this series in a class of its own. Ideal for high school students and advanced grade levels.
Manufacturer:  Motic Swift Line M36044
Catalog No. S80699
Add to cart
Description
Description
OPTICAL SYSTEM
  • Widefield 10x /18 mm eyepiece.
  • Monocular viewing head with 45° inclined eyepieces.
  • DIN standard 4x (0.10 N.A.), 10x (0.25 N.A.), 40xR (0.65 N.A.) and 100xR (1.25 N.A.) achromat objectives. All are parfocalled, parcentered and color-coded.
  • Quad objective turret is ball-bearing mounted for smooth, precise positioning of objectives. Mounted in reverse position to facilitate ease in changing slides.
  • Camera port integrated into head for attachment of optional Moticam camera
  • C-mount lens and 1.5 mm wrench included
ILLUMINATION
  • Variable LED light provides bright, white illumination.
  • LED technology provides cool, energy-efficient light for up to 50,000 hours.
ELECTRICAL
  • 110 V power supply, unit shipped with American plug.
FOCUSING
  • Low position coarse and fine focusing controls.
  • Stage moves up or down, eyepiece and body remain fixed.
  • Slip clutch prevents damage to focusing system.
  • Tension adjustment eliminates stage drift.
  • Safety rack stop prevents damage to slides and objective lenses.
STAGE
  • Large 5-3/4 in. x 4-3/4 in. (143 mm x 120 mm) stage; built-in, low profile, ball-bearing mounted mechanical specimen holder with low-position coaxial controls.
  • Built-in 1.25 Abbe condenser has iris diaphragm.
Specifications
Specifications
M3604-4
Monocular Microscope
40X, 100X, 400X, 1000X
Achromatic