Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 1070722, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-4817140
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent, selective GSK-3 inhibitor

Potent glycogen synthase kinase-3 (GSK-3) inhibitor (Ki = 0.6 nM for GSK-3α and GSK-3β). Displays high selectivity (> 50-fold) for GSK-3 over a panel of other kinases tested, including CDK family members. Decreases phosphorylation of microtubule-associated protein Tau in vitro; protects rat primary cortical neurons against β amyloid and glutamate challenge. Brain penetrant.

Chemical Identifiers

CAS 1384424-80-9
Molecular Formula C17H13F3N4O2
Molecular Weight (g/mol) 362.312
InChI Key VQPBIJGXSXEOCU-UHFFFAOYSA-N
Synonym kinome_2278, kinome_2324, 1-7-methoxyquinolin-4-yl-3-6-trifluoromethyl pyridin-2-yl urea
PubChem CID 49830684
IUPAC Name 1-(7-methoxyquinolin-4-yl)-3-[6-(trifluoromethyl)pyridin-2-yl]urea
SMILES COC1=CC2=NC=CC(=C2C=C1)NC(=O)NC3=CC=CC(=N3)C(F)(F)F

Specifications

Quantity 10 mg
Formula Weight 362.31
Percent Purity >99%
Chemical Name or Material A 1070722
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.