Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A 331440 dihydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1049740-32-0
C22H29Cl2N3O
422.394
XFPYVTSGYYMTFS-GHVWMZMZSA-N
a 331440 dihydrochloride,a-331440 dihydrochloride,a-331440 dihydrochloride hplc , solid,4 inverted exclamation mark-3-3 r-dimethylamino-1-pyrrolidinyl propoxy-1,1-biphenyl-4 inverted exclamation mark r-carbonitrile dihydrochloride,4'-3-3r-3-dimethylamino-1-pyrrolidinyl propoxy-1,1'-biphenyl-4-carbonitrile dihydrochloride,4'-3-3r-3-dimethylamino pyrrolidin-1-yl propoxy-1,1'-biphenyl-4-carbonitrile dihydrochloride
16078942
4-[4-[3-[(3R)-3-(dimethylamino)pyrrolidin-1-yl]propoxy]phenyl]benzonitrile;dihydrochloride
CN(C)C1CCN(C1)CCCOC2=CC=C(C=C2)C3=CC=C(C=C3)C#N.Cl.Cl
This item is not returnable. View return policy
1049740-32-0
C22H29Cl2N3O
422.394
XFPYVTSGYYMTFS-GHVWMZMZSA-N
a 331440 dihydrochloride,a-331440 dihydrochloride,a-331440 dihydrochloride hplc , solid,4 inverted exclamation mark-3-3 r-dimethylamino-1-pyrrolidinyl propoxy-1,1-biphenyl-4 inverted exclamation mark r-carbonitrile dihydrochloride,4'-3-3r-3-dimethylamino-1-pyrrolidinyl propoxy-1,1'-biphenyl-4-carbonitrile dihydrochloride,4'-3-3r-3-dimethylamino pyrrolidin-1-yl propoxy-1,1'-biphenyl-4-carbonitrile dihydrochloride
16078942
4-[4-[3-[(3R)-3-(dimethylamino)pyrrolidin-1-yl]propoxy]phenyl]benzonitrile;dihydrochloride
CN(C)C1CCN(C1)CCCOC2=CC=C(C=C2)C3=CC=C(C=C3)C#N.Cl.Cl

Selective H3 antagonist

High affinity histamine H3 receptor antagonist (Ki values are 21.7 and 22.7 nM for rat and human H3 receptors, respectively). Exhibits selectivity for human H3 over H1, H2 and H4 receptors (Ki values are 2940, 14400 and >10000 nM respectively). Shown to reduce weight in a diet-induced obesity model.
Quantity 50mg
Formula Weight 422.39
Chemical Name or Material A 331440 dihydrochloride

Recommended Storage : Desiccate at RT