Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
A 331440 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity histamine H3 receptor antagonist (Ki values are 21.7 and 22.7 nM for rat and human H3 receptors, respectively). Exhibits selectivity for human H3 over H1, H2 and H4 receptors (Ki values are 2940, 14400 and >10000 nM respectively). Shown to reduce weight in a diet-induced obesity model.
Specifications
Quantity | 10 mg |
Formula Weight | 422.39 |
Chemical Name or Material | A 331440 dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction